Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipic semialdehyde methyl ester, 95%

Catalog No. AC438912500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438912500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438912500 Supplier Thermo Scientific Chemicals Supplier No. 438912500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6654-36-0 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6654-36-0
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
InChI Key FDNFXHCDOASWAY-UHFFFAOYSA-N
Synonym adipic semialdehyde methyl ester, methyl 5-formylvalerate, methyl adipaldehydate, 6-oxo-hexanoic acid methyl ester, 6-oxohexanoic acid methyl ester, hexanoic acid, 6-oxo-, methyl ester, 5-carbomethoxy-1-pentanal, hexanoic acid,6-oxo-,methyl ester, methyl 6-oxo-hexanoate, methyl 5-formylpentanoate
PubChem CID 81164
IUPAC Name methyl 6-oxohexanoate
SMILES COC(=O)CCCCC=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Boiling Point 59°C to 61°C (1.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula OHCCH2CH2CH2CH2CO2CH3
Quantity 250 mg
Formula Weight 144.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Adipic semialdehyde methyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Call a POISON CENTER/doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 229-678-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.