Learn More
ATCC Adipocyte Differentiation Toolkit for Adipose -Derived MSCs and Preadipocytes
Catalog No. 502383027
Encompass
The Adipocyte Differentiation Toolkit for Adipose-derived MSCs and Preadipocytes contains medium and reagents designed to induce adipogenesis in actively proliferating Adipose-Derived Mesenchymal Stem Cells and Preadipocytes with high efficiency, and to support the maturation of derived adipocytes during lipid accumulation.
Catalog No. 50-238-3027 Supplier ATCC Supplier No. PCS500050
The Adipocyte Differentiation Toolkit for Adipose-derived MSCs and Preadipocytes contains medium and reagents designed to induce adipogenesis in actively proliferating Adipose-Derived Mesenchymal Stem Cells and Preadipocytes with high efficiency, and to support the maturation of derived adipocytes during lipid accumulation.