Learn More
Zen Bio Inc ADIPOCYTE MEDIUM
SDP
Catalog No. NC9404747
Encompass
Complete and ready to use culture medium for maintenace of 3T3-L1 adipocytes
Catalog No. NC9404747 Supplier Zen Bio Inc Supplier No. AM1L1
Complete and ready to use culture medium for maintenace of 3T3-L1 adipocytes