Learn More
Promocell GmbH PromoCell Adipocyte Nutrition Medium KIT 500 ml, Contains: C-27431, C-39439
Catalog No. 50306276
Encompass
PromoCell Adipocyte Nutrition Medium is a low-serum (3% V/V) medium developed for the in vitro cultivation of mature adipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Nutrition Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Nutrition Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.03 ml / ml || d-Biotin | 8 μg / ml || Insulin (recombinant human) | 0.5 μg / ml || Dexamethasone | 400 ng / ml |
Catalog No. 50-306-276 Supplier Promocell GmbH Supplier No. C27439
PromoCell Adipocyte Nutrition Medium is a low-serum (3% V/V) medium developed for the in vitro cultivation of mature adipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Nutrition Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Nutrition Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.03 ml / ml || d-Biotin | 8 μg / ml || Insulin (recombinant human) | 0.5 μg / ml || Dexamethasone | 400 ng / ml |