Learn More
Cayman Chemical Adipogenesis Assay Kit
SDP
Catalog No. 502886991
Encompass_Preferred
Cayman’s Adipogenesis Assay provides the reagents required for studying the induction and inhibition of adipogenesis in the established 3T3-L1 model, using the adipogenesis induction procedure. This kit can also be used to screen drug candidates involved in adipogenesis. The classic Oil Red O staining for lipid droplets is used in this kit as an indicator of the degree of adipogenesis, and can be quantified with a plate reader after the dye is conveniently extracted from the lipid droplet.
Catalog No. 50-288-6991 Supplier Cayman Chemical Supplier No. 100069081 EA
Add to Cart
Add to Cart
Cayman’s Adipogenesis Assay provides the reagents required for studying the induction and inhibition of adipogenesis in the established 3T3-L1 model, using the adipogenesis induction procedure. This kit can also be used to screen drug candidates involved in adipogenesis. The classic Oil Red O staining for lipid droplets is used in this kit as an indicator of the degree of adipogenesis, and can be quantified with a plate reader after the dye is conveniently extracted from the lipid droplet.