Learn More
STEMCELL Technologies MesenCult™ Adipogenic Differentiation Kit (Mouse), Size: 1 Kit
SDP
Catalog No. NC1611467
Encompass
MesenCult™ Adipogenic Differentiation Kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs), adipose tissue-derived MSCs (ADSCs), and mouse embryonic fibroblasts (MEFs) into cells of the adipogenic lineage.NOTE: MesenCult™ Adipogenic Differentiation Medium must be supplemented with L-Glutamine (Catalog #07100).
Catalog No. NC1611467 Supplier STEMCELL Technologies Supplier No. 05507
MesenCult™ Adipogenic Differentiation Kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs), adipose tissue-derived MSCs (ADSCs), and mouse embryonic fibroblasts (MEFs) into cells of the adipogenic lineage.NOTE: MesenCult™ Adipogenic Differentiation Medium must be supplemented with L-Glutamine (Catalog #07100).