Learn More
Lonza Walkersville Adipogenic Induction Medium
Supplier:  Lonza Walkersville PT3102B
Encompass
Adipogenic Induction Medium is a component of the hMSC Adipogenic Differentiation BulletKit and is availalbe separately Stem cells possess the unique ability to differentiate into multiple lineages of cells including adipocytes Lonza s hMSC Adipogenic Differentiation BulletKit Medium is a serum containing media system designed to induce adipogenic differentiation of human bone marrow derived mesenchymal stem cells into mature functionally active adipocytes in as few as 19 days Differentiation can then be validated by a variety of methods including the use of Lonza s AdipoRed Adipogenic Assay Reagent which provides a quantitative value related to the amount of adipogenic differentiation
Catalog No. 50-004-96436