Learn More
OriGene Adiponectin Receptor 1 (ADIPOR1) CytoSection, 25 slides
SDP
Catalog No. NC3786275
Encompass_Preferred
Transient overexpression of ADIPOR1, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3786275 Supplier OriGene Supplier No. TS403907P5
Transient overexpression of ADIPOR1, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack