Learn More
Enzo Life Sciences Adiponectin polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905714100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human Adiponectin. UniProt: Q15848. Species Reactivity: Human, Mouse, Rat. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (1µg/ml). Western Blot (1µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-658-465