Learn More
Bioss Adiponectin C-terminal region
SDP
Catalog No. 5000065179
Encompass_Preferred
Adiponectin (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M577Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65179 Supplier Bioss Supplier No. BSM70326M
Adiponectin (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M577Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol