Learn More
Medchemexpress LLC HY-P7126 10ug Medchemexpress, Adiponectin (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501934350
Encompass_Preferred
Medchemexpress, HY-P7126 10ug Adiponectin (CHO-expressed), Human CAS: Recombinant Human Adiponectin (CHO-expressed) could increase cell sensitivity to insulin and can be used as a potential protein for treating diabetic tendinopathy Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4350 Supplier Medchemexpress LLC Supplier No. HYP712610UG
Medchemexpress, HY-P7126 10ug Adiponectin (CHO-expressed), Human CAS: Recombinant Human Adiponectin (CHO-expressed) could increase cell sensitivity to insulin and can be used as a potential protein for treating diabetic tendinopathy Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.