Learn More
Crystal Chem Inc Adiponectin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80571
Encompass
The Adiponectin ELISA is used to quantify levels of human adiponectin in serum and plasma using only a 10 µL sample volume. Highlights: Kits use only 10 µL sample. Dynamic range of 2 - 100 ng/mL. High sensitivity of 0.3 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7738