Learn More
AdipoGen Adiponectin (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0030C050
Encompass_Preferred
Protein. Human adiponectin (aa 16-244) is fused at the N-terminus to a FLAG-tag. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). The purified Adiponectin (human) (rec.) (Prod. No. AG-40B-0030) is a mixture of high molecular weight (HMW) and hexameric active species.
Catalog No. 50-205-3104