Learn More
Enzo Life Sciences Adiponectin (human), ELISA Kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850377KI01
Encompass_Preferred
The Adiponectin (human), ELISA kit is a colorimetric, immunometric immunoassay kit with results in <3 hours. Alternative name: ACRP30, ADIPOQ, GBP28, Adipocyte C1q and collagen domain-containing protein, Adipocyte complement-related 30 kDa protein, Gelatin-binding protein, ACDC, APM1. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-0165