Learn More
AdipoGen Adiponectin human pAb
SDP
Supplier:  AdipoGen AG25A0003C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ACRP30 was identified as a novel adipocyte-specific synthesized and secreted protein with structural resemblance to complement factor C1q. Like adipsin, ACRP30 secretion is induced ~10-fold during adipocyte differentiation. Plasma levels are reduced in obese humans, and low levels are associated with insulin-resistance. Treatment of db/db mice with TZD increased ACRP30 levels.
Catalog No. 50-256-3309