Learn More
Medchemexpress LLC HY-P7129 10ug Medchemexpress, Adiponectin, Mouse CAS: Purity:>98%
SDP
Catalog No. 502083109
Encompass_Preferred
Medchemexpress, HY-P7129 10ug Adiponectin, Mouse CAS: Recombinant Mouse Adiponectin suppresses endothelial inflammatory response and vascular smooth muscle cell proliferation, as well as macrophage-to-foam cell transformation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3109 Supplier Medchemexpress LLC Supplier No. HYP712910UG
Medchemexpress, HY-P7129 10ug Adiponectin, Mouse CAS: Recombinant Mouse Adiponectin suppresses endothelial inflammatory response and vascular smooth muscle cell proliferation, as well as macrophage-to-foam cell transformation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.