Learn More
Enzo Life Sciences Adiponectin (mouse), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522059C050
Encompass_Preferred
Alternative name: Adipocyte complement related protein of 30kDa, ACRP30, AdipoQ, apM1, GBP28. Concentration: 0.5mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. Mouse adiponectin (aa 18-247) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Long Term Storage: -20°C.
Catalog No. 50-200-9397