Learn More
AdipoGen Adiponectin (mouse) ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45A0004YEKKI01
Encompass_Preferred
ELISA Assay. Detects mouse adiponectin. Does not cross-react with human adiponectin, rat adiponectin, mouse resistin, mouse RELM-beta or mouse leptin. Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum. Range: 0.125 to 8ng/ml. Sensitivity: 50pg/ml. Adiponectin is a promising biomarker of insulin resistance and type 2 diabetes mellitus (T2DM) but also as a potential target for management of the metabolic syndrome. It is a very robust marker that is not prone to degradation or acute inflammatory challenges, is present in relatively high concentrations in the peripheral circulation, and can be collected by a variety of methods. Adiponectin exerts anti-atherogenic and anti-inflammatory properties and may be important as a biomarker for obesity-related cardiovascular disease (CVD). Adiponectin serum level was also described as a good biomarker of colorectal adenoma.
Catalog No. 50-205-3306