Learn More
AdipoGen Adiponectin rat ELISA Kit
SDP
Supplier:  AdipoGen AG45B0026KI01
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Detects rat adiponectin. Does cross-react with mouse adiponectin or human adiponectin. Assay Type Sandwich. Detection Type Colorimetric. Sample Type Cell Culture Supernatant, Plasma, Serum. Range 0.0625 to 4ng/ml. Sensitivity 50pg/ml. Adiponectin is a promising biomarker of insulin resistance and type 2 diabetes mellitus T2DM but also as a potential target for management of the metabolic syndrome. It is a very robust marker that is not prone to degradation or acute inflammatory challenges, is present in relatively high concentrations in the peripheral circulation, and can be collected by a variety of methods. The benefits of using adiponectin assays in clinical settings include, a prediction of risk of diabetes and metabolic status and b providing a tool to monitor metabolic improvements. Adiponectin exerts anti-atherogenic and anti-inflammatory properties and may be important as a biomarker for obesity-related cardiovascular disease CVD.
Catalog No. 50-256-2733