Learn More
Alpco Ltd Rat Adiponectin ELISA
SDP
Supplier:  Alpco Ltd 22ADPRTE01
Encompass_Preferred
The Rat Adiponectin ELISA kit is a tool for the quantification of rat adiponectin in serum and plasma samples. Utilizing a dual-monoclonal antibody sandwich format, bi-level control set, and 96-well microplate comprising removable strips, a single Rat Adiponectin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if the entire kit is not needed at one time.
Analytical sensitivity
0.08 ng/mL
Dynamic range
0.25-10 ng/mL
Low sample volume
5 µL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3533