Learn More
AdipoGen Adiponectin (rat) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40A0004YC050
Encompass_Preferred
Recombinant Proteins. Rat adiponectin (aa 16-244) is fused at the N-terminus to a FLAG-tag. Source: HEK 293 cells. Lyophilized. Contains 30mM Tris-Cl, pH 8.0 + 100mM NaCl. Endotoxin levels: <0.01EU/µg purified protein (LAL test).. ACRP30 was identified as a novel adipocyte-specific synthesized and secreted protein with structural resemblance to complement factor C1q. Like adipsin, ACRP30 secretion is induced ~10-fold during adipocyte differentiation. Plasma levels are reduced in obese humans and low levels are associated with insulin-resistance. Treatment of db/db mice with TZD increased ACRP30 levels.
Catalog No. 50-205-3059