Learn More
Medchemexpress LLC Adiponectin receptor protein 1 Antibody (YA2700) | 100 UL
SDP
Catalog No. 5000443642
Encompass_Preferred
The MedChemExpress Adiponectin receptor protein 1 Antibody (YA2700) is a rabbit-derived antibody supplied as a liquid, primarily for research applications. This non-conjugated antibody has a molecular weight of 43 kDa and is suitable for various laboratory uses.
  • Reacts with human, mouse, and rat samples
  • Stable for 1 year when stored at -20°C
  • Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Recommended for Western Blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC)
  • Shipped with blue ice
  • Avoid repeated freeze/thaw cycles
Catalog No. 50-004-43642 Supplier Medchemexpress LLC Supplier No. HYP82955100UL
The MedChemExpress Adiponectin receptor protein 1 Antibody (YA2700) is a rabbit-derived antibody supplied as a liquid, primarily for research applications. This non-conjugated antibody has a molecular weight of 43 kDa and is suitable for various laboratory uses.
  • Reacts with human, mouse, and rat samples
  • Stable for 1 year when stored at -20°C
  • Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Recommended for Western Blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC)
  • Shipped with blue ice
  • Avoid repeated freeze/thaw cycles