Learn More
AdipoGen anti-Adiponectin (rat), mAb (RADI 264) (50 µg)
SDP
Supplier:  AdipoGen AG20A0036C050
Encompass_Preferred
Monoclonal Antibody. Recognizes rat adiponectin. Detects a band of ~30kDa by Western blot. Does not cross-react with human or mouse adiponectin. Isotype: Mouse IgG1kappa. Clone: RADI 264. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ACRP30 was identified as a novel adipocyte-specific synthesized and secreted protein with structural resemblance to complement factor C1q. Like adipsin, ACRP30 secretion is induced ~10-fold during adipocyte differentiation. Plasma levels are reduced in obese humans, and low levels are associated with insulin-resistance. Treatment of db/db mice with TZD increased ACRP30 levels.
Catalog No. 50-205-2825