Learn More
Novus Biologicals™ Adiponutrin/PNPLA3 Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 80339 |
| Species | Mouse |
| Purity | >50%, by HPLC |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Format | Lyophilized |
| Formulation | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-00227. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Acylglycerol O-acyltransferase, adiponutrin, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3 |
| Gene Symbol | PNPLA3 |
| Quantity | 0.1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.