Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP527925UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Human. BioActivity: gACRP (globular adiponectin) activity was confirmed by measuring inhibition of cytotoxicity of M1 cells. Typical ED50 values for our recombinant adiponectin in this assay are in the range of 3 ug/ml. AA Sequence: MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN. Buffer: 10 mM Tris, 0.75 mM DTT, pH 8.0. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 5.00 EUs/ug as determined by the LAL method. Gene Symbol: ADIPOQ. Gene ID: 9370. Uniprot: Q15848.
Catalog No. 50-188-5754