Learn More
Aviva Systems Biology Corp ADIPOR1 Antibody - N-terminal region (ARP63340_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63340_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADIPOR1. It was validated on Western Blot. . Alternate names: ACDCR1, CGI-45, CGI45, FLJ25385, FLJ42464, PAQR1, TESBP1AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-942