Learn More
Aviva Systems Biology Corp ADIPOR2 Antibody - N-terminal region (ARP60818_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60818_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADIPOR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ADIPOR2. Alternate names: ACDCR2, FLJ21432, MGC4640, PAQR2ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rabbit
Catalog No. 50-287-179