Learn More
Enzo Life Sciences Adiponectin receptor 2 (mouse) polyclonal antibody (200µg)
SDP
Catalog No. 502008144
Encompass_Preferred
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 11-40 (C11TRTPEPDIRLRKGHQLDDTRGSNNDNYQG40) of mouse adiponectin receptor 2 (AdipoR2). Species reactivity: Mouse. Applications: ICC, IHC (PS), WB. Purity: Epitope affinity-purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8144 Supplier Enzo Life Sciences Supplier No. ALX210647C200
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 11-40 (C11TRTPEPDIRLRKGHQLDDTRGSNNDNYQG40) of mouse adiponectin receptor 2 (AdipoR2). Species reactivity: Mouse. Applications: ICC, IHC (PS), WB. Purity: Epitope affinity-purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.