Learn More
Lonza Walkersville AdipoRed Assay Reagent, 20 mL
Catalog No. NC9049267
Encompass
The AdipoRed Assay Reagent is designed for assessing the effect of compounds on the differentiation of preadipocytes or on lipid utilization in mature adipocytes. The lipophilic AdipoRed Assay Reagent specifically partitions into the fat droplets of differentiated adipocytes and fluoresces at 572 nm.This objective, high-throughput, homogeneous fluorescence-based assay quantifies the accumulation of intracellular triglycerides and provides significant advantages to drug discovery efforts in the field of obesity and diabetes.Benefits:Convenient - Simply remove cell culture medium, add premixed reagent and read in a standard fluorometerFast- Process an entire 96-well or 384-well plate in as little as 20 minutesEffective - Provides objective, high throughput measurement of the accumulation of intracellular triglycerides, with high signal to noise ratiosContent:5 x AdipoRed Assay Reagent, 4 mL
Catalog No. NC9049267 Supplier Lonza Walkersville Supplier No. PT7009
The AdipoRed Assay Reagent is designed for assessing the effect of compounds on the differentiation of preadipocytes or on lipid utilization in mature adipocytes. The lipophilic AdipoRed Assay Reagent specifically partitions into the fat droplets of differentiated adipocytes and fluoresces at 572 nm.This objective, high-throughput, homogeneous fluorescence-based assay quantifies the accumulation of intracellular triglycerides and provides significant advantages to drug discovery efforts in the field of obesity and diabetes.Benefits:Convenient - Simply remove cell culture medium, add premixed reagent and read in a standard fluorometerFast- Process an entire 96-well or 384-well plate in as little as 20 minutesEffective - Provides objective, high throughput measurement of the accumulation of intracellular triglycerides, with high signal to noise ratiosContent:5 x AdipoRed Assay Reagent, 4 mL