Learn More
AdipoGen AdipoRon, 924416-43-3, MFCD08271060, 10 mg
SDP
Supplier:  AdipoGen AGCR10154M010
Encompass_Preferred
MF: C27H28N2O3, MW: 428.5, ≥98%, Appearance: White solid. Orally-active adiponectin receptor (AdipoR) agonist. Binds to AdipoR1 and AdipoR2 at low μm concentration. Activates 5'-adenosine monophosphate-activated protein kinase (AMPK) in cultured mammalian cells. Improves diabetes, glucose and lipid metabolism and insulin sensitivity in cultured cells and in mice by AdipoR-dependent mechanisms.
Catalog No. 50-101-6971