Learn More
Apexbio Technology LLC AdipoRon, 10mg. Cas: 924416-43-3 MFCD: MFCD08271060. AdipoR1 and AdipoR2 agonist, first orally active
SDP
Supplier:  Apexbio Technology LLC B187910
Encompass_Preferred
AdipoRon is an agonist of AdipoR1 and AdipoR2 with Kd values of 1.8 and 3.1 M, respectively [1]. Adiponectin receptor (AdipoR) is a receptor for adiponectin, which is a protein and plays an important role in regulating glucose levels and fatty acid breakdown. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5289