Learn More
Apexbio Technology LLC AdipoRon 924416-43-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063722
Encompass_Preferred
AdipoRon (CAS 924416-43-3) is an agonist of adiponectin receptors AdipoR1 and AdipoR2 displaying binding affinities (Kd) of 1 8 M and 3 1 M respectively Upon receptor activation it promotes phosphorylation of AMPK via AdipoR1 and upregulates expression of PGC-1 improving mitochondrial biogenesis in muscle cell models In vivo studies show that AdipoRon administration stimulates AMPK and PPAR- signaling in liver and muscle tissues subsequently reducing circulating glucose insulin triglycerides and inflammatory cytokines This compound is employed in metabolic and cardiovascular disease research especially regarding insulin resistance dyslipidemia obesity-related inflammation and ischemia-induced myocardial damage
Catalog No. 50-000-63722 Supplier Apexbio Technology LLC Supplier No. B18795.1
AdipoRon (CAS 924416-43-3) is an agonist of adiponectin receptors AdipoR1 and AdipoR2 displaying binding affinities (Kd) of 1 8 M and 3 1 M respectively Upon receptor activation it promotes phosphorylation of AMPK via AdipoR1 and upregulates expression of PGC-1 improving mitochondrial biogenesis in muscle cell models In vivo studies show that AdipoRon administration stimulates AMPK and PPAR- signaling in liver and muscle tissues subsequently reducing circulating glucose insulin triglycerides and inflammatory cytokines This compound is employed in metabolic and cardiovascular disease research especially regarding insulin resistance dyslipidemia obesity-related inflammation and ischemia-induced myocardial damage