Learn More
Apexbio Technology LLC AdipoRon, 924416-43-3, MFCD08271060, 50 mg
SDP
Supplier:  Apexbio Technology LLC B187950
Encompass_Preferred
MF: C27H28N2O3, MW: 428.52, Solubility: 21.45mg/mL in DMSO, Chemical Name: 2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide. AdipoRon is an agonist of AdipoR1 and AdipoR2 with Kd values of 1.8 and 3.1 uM, respectively. Adiponectin receptor (AdipoR) is a receptor for adiponectin, which is a protein and plays an important role in regulating glucose levels and fatty acid breakdown.
Catalog No. 50-101-2739