Learn More
Enzo Life Sciences AdipoRon (5mg). CAS: 924416-43-3
SDP
Catalog No. 502013374
Encompass_Preferred
AdipoR agonist that binds to both AdipoR1 and AdipoR2. Alternative name: 2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (86mg/ml) or ethanol (50mg/ml). Slightly soluble in water (<1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-3374 Supplier Enzo Life Sciences Supplier No. ENZCHM1010005
AdipoR agonist that binds to both AdipoR1 and AdipoR2. Alternative name: 2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (86mg/ml) or ethanol (50mg/ml). Slightly soluble in water (<1mg/ml). Long Term Storage: -20°C.