Learn More
AdipoGen AdipoRon . hydrochloride (water soluble) (50 mg)
SDP
Supplier:  AdipoGen AGCR10156M050
Encompass_Preferred
Chemical. CAS: 924416-43-3 (free base). Formula: C27H28N2O3 . HCl . H2O. MW: 428.5 . 36.5 . 18.0. Orally-active adiponectin receptor (AdipoR) agonist. Binds to AdipoR1 and AdipoR2 at low µm concentration. Activates 5'-adenosine monophosphate-activated protein kinase (AMPK) in cultured mammalian cells. Activates peroxisome proliferator-activated receptor gamma coactivator 1-alpha (PGC1alpha) which boosts mitochondrial proliferation and energy metabolism. Improves diabetes, glucose and lipid metabolism and insulin sensitivity in cultured cells and in mice by AdipoR-dependent mechanisms. Rescued the shortened lifespan of db/db mice (AdipoRs KO) on high-fat diet. Reduces expression levels of genes encoding inflammatory cytokines such as TNF-alpha, IL-6 and CCL-2 in WAT of WT mice but not db/db mice.
Catalog No. 50-205-3857