Learn More
Invent Biotechnologies Inc Minute™ Total Protein Extraction Kit for Adipose Tissues/Cultured Adipocytes, 20 preps, spin-column-based, detergent-free extraction, high yield 2-3 mg/ml, 50-80 mg adipose tissue
Catalog No. NC1471072
Encompass
Minute™ Total Protein Extraction Kit for Adipose Tissues/Cultured Adipocytes, 20 preps, spin-column-based, detergent-free extraction, high yield 2-3 mg/ml, 50-80 mg adipose tissue
Catalog No. NC1471072 Supplier Invent Biotechnologies Inc Supplier No. AT022
Minute™ Total Protein Extraction Kit for Adipose Tissues/Cultured Adipocytes, 20 preps, spin-column-based, detergent-free extraction, high yield 2-3 mg/ml, 50-80 mg adipose tissue