Learn More
Invent Biotechnologies Inc Minute™ Adipose Tissue Fractionation Kit - Spin Column-Based (20 Preps)
Catalog No. 501978734
Encompass
On average, total cellular proteins account for less than 2% of adipose tissue. Adipose tissue, especially white adipose tissue, has been recognized as an important endocrine and inflammation organ in addition to its energy storage function. Fractionation and analysis of proteins from adipose tissue are critical for understanding many physiological/pathological conditions. However, fractionation of adipose tissue is challenging due to its high lipid and low protein contents. The water-oil emulsion present in biological samples is notoriously difficult to separate, but we have developed a cutting-edge technology to deal with this issue: a porous filter with unique surface property and predefined pore size coupling that has a specially formulated detergent-free fractionation buffer.
Catalog No. 50-197-8734 Supplier Invent Biotechnologies Inc Supplier No. AF023
On average, total cellular proteins account for less than 2% of adipose tissue. Adipose tissue, especially white adipose tissue, has been recognized as an important endocrine and inflammation organ in addition to its energy storage function. Fractionation and analysis of proteins from adipose tissue are critical for understanding many physiological/pathological conditions. However, fractionation of adipose tissue is challenging due to its high lipid and low protein contents. The water-oil emulsion present in biological samples is notoriously difficult to separate, but we have developed a cutting-edge technology to deal with this issue: a porous filter with unique surface property and predefined pore size coupling that has a specially formulated detergent-free fractionation buffer.