Learn More
ATCC Adipose-Derived Mesenchymal Stem Cells; Normal, Human
Supplier:  ATCC PCS500011
Encompass
Normal Human Adipose-Derived Mesenchymal Stem Cells are fibroblast-like cells that, when grown in Mesenchymal Stem Cell Basal Media supplemented with Mesenchymal Stem Cell Growth Kit Low serum components, provide an ideal cell system to propagate mesenchymal stem cells in low serum (2% FBS) conditions. When maintained under optimal growth conditions, ATCC Normal Human Adipose-Derived Mesenchymal Stem Cells have been shown to be multipotent, capable of differentiating down the adipogenic, osteogenic, and chondrogenic lineages. The cells are cryopreserved at the second passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements, and reagents are quality tested together to guarantee optimum performance and reliability.
Catalog No. 50-238-2303
 Restricted Item