Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Adipoyl Chloride, Hexane Solution
SCI_ED

Catalog No. S25125A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25125A 250 mL Solution
S25125 100 mL Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25125A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Nylon production

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 111-50-2 , 11-54-3
Molecular Formula C6H8Cl2O2
InChI Key PWAXUOGZOSVGBO-UHFFFAOYSA-N
IUPAC Name hexanedioyl dichloride

Specifications

Color Colorless
Density 0.66 to 0.69 g/mL
Assay Percent Range <3.65% w/v
Packaging Solution
Quantity 250 mL
Synonym 2,5-pyrimidinedicarboxylic acid, 2,5-dimethyl ester, 2,5-dimethylpyrimidine-2,5-dicarboxylate, 2,5-dimethyl pyrimidine-2,5-dicarboxylate
SMILES ClC(=O)CCCCC(Cl)=O
Molecular Weight (g/mol) 183.03
PubChem CID 14470811
Formula Weight Mixture
Chemical Name or Material Adipoyl Chloride, Hexane Solution
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife One year
DOTInformation DOT Class 3 (8), II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.