Learn More
Cayman Chemical AdIpoylCoenzyme AsodIM sa 10mg
SDP
Catalog No. 502941759
Encompass_Preferred
A medium-chain acyl CoA; an intermediate in the peroxisomal β-oxidation of dicarboxylic acids; an intermediate in the biosynthesis of adipic acid in genetically modified E. coli
Catalog No. 50-294-1759 Supplier Cayman Chemical Supplier No. 2785810 MG
A medium-chain acyl CoA; an intermediate in the peroxisomal β-oxidation of dicarboxylic acids; an intermediate in the biosynthesis of adipic acid in genetically modified E. coli