Learn More
MSC Miller Adjustable Barrel Mount Sleeve, 24 in. Capacity, Anodized Aluminum, 1 ea.
Catalog No. 5000509478
Encompass
This adjustable barrel-mount sleeve is engineered for shoring, wall, and parapet applications that require frequent setup over varying wall thicknesses. Constructed from durable, welded aluminum, it provides a reliable mounting solution for confined space systems.
  • Accommodates wall thicknesses up to 610 mm (24 in.)
  • Features 450 lbs. (204.1 kg) maximum working load capacity
  • Includes eight adjustment pins for secure configuration
  • Incorporates a PVC sleeve liner conforming to ASTM D-1785
  • Designed for use with approved masts and accessories
  • Meets OSHA, ANSI, and CSA safety standards
Catalog No. 50-005-09478 Supplier MSC Supplier No. 61950481
This adjustable barrel-mount sleeve is engineered for shoring, wall, and parapet applications that require frequent setup over varying wall thicknesses. Constructed from durable, welded aluminum, it provides a reliable mounting solution for confined space systems.
  • Accommodates wall thicknesses up to 610 mm (24 in.)
  • Features 450 lbs. (204.1 kg) maximum working load capacity
  • Includes eight adjustment pins for secure configuration
  • Incorporates a PVC sleeve liner conforming to ASTM D-1785
  • Designed for use with approved masts and accessories
  • Meets OSHA, ANSI, and CSA safety standards