Learn More
Alzet ALZETTM Mouse Jugular Catheter with Adjustable Length: Optimized for Jugular Delivery from any ALZET Osmotic Pump
SDP
Catalog No. NC9009177
Encompass
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous IV delivery of test agents in mice. EtO sterilized and ready for immediate use.Catheter Material: PolyurethaneSpecifications: 8.9 cm total length; distal tip 1 Fr; internal volume ~18 μlOptimum Animal Size: 30+ grams (insertion marks for adaptation to smaller animals)Features & Benefits:•ALZET connection can be trimmed to achieve shorter lengths•Catheter tip optimum for use with other small vessels (e.g., femoral, carotid, etc.)•Retention beads at 1.3 and 3.5 cm from the tip facilitate anchoring to tissue•Marked at 0.9 and 1.1 cm to simplify adaptation for animals weighing <25 g and 25-30 g respectively
Catalog No. NC9009177 Supplier Alzet Supplier No. 0007701
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous IV delivery of test agents in mice. EtO sterilized and ready for immediate use.Catheter Material: PolyurethaneSpecifications: 8.9 cm total length; distal tip 1 Fr; internal volume ~18 μlOptimum Animal Size: 30+ grams (insertion marks for adaptation to smaller animals)Features & Benefits:•ALZET connection can be trimmed to achieve shorter lengths•Catheter tip optimum for use with other small vessels (e.g., femoral, carotid, etc.)•Retention beads at 1.3 and 3.5 cm from the tip facilitate anchoring to tissue•Marked at 0.9 and 1.1 cm to simplify adaptation for animals weighing <25 g and 25-30 g respectively