Learn More
MSC Fantech Exhaust Grille, Sheet Steel, Adjustable, 8 in., 1 ea.
Catalog No. 5000506967
Encompass
This adjustable metal exhaust grille is designed for efficient intake or exhaust applications in both ceiling and wall installations. The unit is constructed from durable sheet steel with a white powder-coated finish and includes a galvanized mounting collar with a nailing strip for direct duct installation.
  • Designed for exhaust or supply air systems
  • Adjustable pressure and air volume via a lockable central rotating cone
  • Constructed from sheet steel with white powder-coated finish (RAL 9010-80)
  • Includes galvanized mounting collar with nailing strip
  • Suitable for wall or ceiling mounting
  • Duct size 8 in.
Catalog No. 50-005-06967 Supplier MSC Supplier No. 72097090
This adjustable metal exhaust grille is designed for efficient intake or exhaust applications in both ceiling and wall installations. The unit is constructed from durable sheet steel with a white powder-coated finish and includes a galvanized mounting collar with a nailing strip for direct duct installation.
  • Designed for exhaust or supply air systems
  • Adjustable pressure and air volume via a lockable central rotating cone
  • Constructed from sheet steel with white powder-coated finish (RAL 9010-80)
  • Includes galvanized mounting collar with nailing strip
  • Suitable for wall or ceiling mounting
  • Duct size 8 in.