Learn More
MSC ADJ OFFSET DAVIT MAST
Catalog No. 5000803480
Encompass
The 3M DBI-SALA Confined Space One-Piece Adjustable Offset Davit Mast is a durable, high-strength component designed for secure fall protection and successful rescue and retrieval in confined spaces. It meets OSHA confined space regulations and features an easily adjustable telescoping design and strong aluminum construction, making it resistant to weather and rugged use.
  • Integrated 5,000 lb. (22 kN) mast anchorage point
  • Rated for fall arrest in different davit offset mast positions
  • Adjustable vertical height
  • For use with portable and permanent bases
  • Durable, corrosion-resistant aluminum construction
  • Telescoping pole for easy adjustment
  • Pins connect quickly for fast set-up and breakdown
Catalog No. 50-008-03480 Supplier MSC Supplier No. 51377885
The 3M DBI-SALA Confined Space One-Piece Adjustable Offset Davit Mast is a durable, high-strength component designed for secure fall protection and successful rescue and retrieval in confined spaces. It meets OSHA confined space regulations and features an easily adjustable telescoping design and strong aluminum construction, making it resistant to weather and rugged use.
  • Integrated 5,000 lb. (22 kN) mast anchorage point
  • Rated for fall arrest in different davit offset mast positions
  • Adjustable vertical height
  • For use with portable and permanent bases
  • Durable, corrosion-resistant aluminum construction
  • Telescoping pole for easy adjustment
  • Pins connect quickly for fast set-up and breakdown