Learn More
Bellco Glass, Inc. Adj. Paddle Blades, 6-8L
SDP
Catalog No. 501945023
Encompass_Preferred
Adjustable PTFE Microcarrier paddle blades only , 6-8L Paddle dimensions: 1 1/2" x 6 3/8". MADE IN THE USA.
Catalog No. 50-194-5023 Supplier Bellco Glass, Inc. Supplier No. 196540008
Adjustable PTFE Microcarrier paddle blades only , 6-8L Paddle dimensions: 1 1/2" x 6 3/8". MADE IN THE USA.