Learn More
Grainger Adj Paint Roll Frame 12 -18 Roll L 18 L
Catalog No. 502748949
Encompass_Preferred
The Premier Adjustable Paint Roller Frame is a 9-inch roller frame that extends from 9 to 18 inches. It has a threaded handle that allows you to use extension poles. The frame is made of plastic with stainless steel bearings. It accepts all 9-inch roller covers and works with all paints and stains. The adjustable frame allows you to cut in around edges and paint large surfaces without changing tools.
Catalog No. 50-274-8949 Supplier Grainger Supplier No. 14C550
The Premier Adjustable Paint Roller Frame is a 9-inch roller frame that extends from 9 to 18 inches. It has a threaded handle that allows you to use extension poles. The frame is made of plastic with stainless steel bearings. It accepts all 9-inch roller covers and works with all paints and stains. The adjustable frame allows you to cut in around edges and paint large surfaces without changing tools.