Learn More
Bellco Glass, Inc. Adj.Sample 1/8 Pipe Assembly , 15L Bioreactor
SDP
Catalog No. 501946495
Encompass_Preferred
Adjusted Sample 1/8” Pipe Assembly Hi -Torque Over Head Drive Adapter is used in conjunction with the High-Torque OHD Motor to drive the impeller assembly for mixing within a vessel. Used with 100mm cap assembly. --316L SST design with polyester/SST bearing --Includes two (2) 1/4” NPT female plugged ports for optimal fittings --Dimensions 3 3/4” l x 3 3/4” w x t 1/2” tall (including dust shield coupling)--Designed to fit with OHD high torque setups --Adapts all bellco PTFE impeller assemblies or SST impeller assemblies. MADE IN THE USA
Catalog No. 50-194-6495 Supplier Bellco Glass, Inc. Supplier No. 790915114
Adjusted Sample 1/8” Pipe Assembly Hi -Torque Over Head Drive Adapter is used in conjunction with the High-Torque OHD Motor to drive the impeller assembly for mixing within a vessel. Used with 100mm cap assembly. --316L SST design with polyester/SST bearing --Includes two (2) 1/4” NPT female plugged ports for optimal fittings --Dimensions 3 3/4” l x 3 3/4” w x t 1/2” tall (including dust shield coupling)--Designed to fit with OHD high torque setups --Adapts all bellco PTFE impeller assemblies or SST impeller assemblies. MADE IN THE USA