Learn More
MSC Gilmour Handheld Sprayer, 32 oz, Plastic Tank, 1 ea
Catalog No. 502867271
Encompass
The Gilmour handheld sprayer is designed for garden and pump applications. It features a plastic tank and synthetic rubber seals, and is not chemical safe. This unit includes a nozzle, quick connectors, and a deflector jet.
  • Not chemical safe
  • Plastic tank construction
  • Synthetic rubber seals and gaskets
  • No hose included
  • Includes nozzle, quick connectors, and deflector jet
  • Compatible with cleaners
  • Volume capacity of 32 oz
  • Plastic wand
  • Funnel style mouth opening
  • Height of 10-1/2 in
Catalog No. 50-286-7271 Supplier MSC Supplier No. 16814261
The Gilmour handheld sprayer is designed for garden and pump applications. It features a plastic tank and synthetic rubber seals, and is not chemical safe. This unit includes a nozzle, quick connectors, and a deflector jet.
  • Not chemical safe
  • Plastic tank construction
  • Synthetic rubber seals and gaskets
  • No hose included
  • Includes nozzle, quick connectors, and deflector jet
  • Compatible with cleaners
  • Volume capacity of 32 oz
  • Plastic wand
  • Funnel style mouth opening
  • Height of 10-1/2 in