Learn More
McMaster-Carr Torque Wrench/Drivers with NIST Certificate
Catalog No. NC2224447
Encompass
The McMaster-Carr 1/4" Drive Click Torque Wrench is a precision torque wrench that provides accurate torque for tightening fasteners. It has a measurement range of 10 to 100 in-lbs and comes with a NIST-traceable certificate verifying its calibration accuracy. The wrench has a reversible ratchet head that allows tightening and loosening, and a knurled grip for comfort. It features a click-type design that makes an audible click when the preset torque value is reached. The torque wrench has an accuracy of ±4% clockwise and ±6% counterclockwise. It is made from chrome vanadium steel and weighs 0.60 lbs. The torque wrench comes with a plastic storage case.
This item is not returnable. View return policy
Catalog No. NC2224447 Supplier McMaster-Carr Supplier No. 85555A311
The McMaster-Carr 1/4" Drive Click Torque Wrench is a precision torque wrench that provides accurate torque for tightening fasteners. It has a measurement range of 10 to 100 in-lbs and comes with a NIST-traceable certificate verifying its calibration accuracy. The wrench has a reversible ratchet head that allows tightening and loosening, and a knurled grip for comfort. It features a click-type design that makes an audible click when the preset torque value is reached. The torque wrench has an accuracy of ±4% clockwise and ±6% counterclockwise. It is made from chrome vanadium steel and weighs 0.60 lbs. The torque wrench comes with a plastic storage case.