Learn More
MSC Miller Two-Piece Adjustable Upper Mast, Aluminum, 12 in. to 29 in. Offset, 1 ea
Catalog No. 5000509863
Encompass
This two-piece adjustable upper mast system is engineered for confined space entry, rescue, and personnel or material handling. Designed for height-restrictive applications, the system provides flexible configuration options and allows for the mounting of multiple winches and self-retracting lifeline systems.
  • Adjustable offset of 12 in. to 29 in.
  • Lightweight aluminum construction features 6061-T6, 5052-H32, and 7075-T6 alloys
  • Tool-less design enables breakdown into manageable components for easy storage, transport, and setup
  • Safety yellow powder coat finish
  • Includes 304 stainless steel block anchor and zinc-plated Grade 5 hardware
  • Rated capacity of 450 lbs
  • Meets all applicable OSHA, ANSI, and CSA standards
Catalog No. 50-005-09863 Supplier MSC Supplier No. 59210724
This two-piece adjustable upper mast system is engineered for confined space entry, rescue, and personnel or material handling. Designed for height-restrictive applications, the system provides flexible configuration options and allows for the mounting of multiple winches and self-retracting lifeline systems.
  • Adjustable offset of 12 in. to 29 in.
  • Lightweight aluminum construction features 6061-T6, 5052-H32, and 7075-T6 alloys
  • Tool-less design enables breakdown into manageable components for easy storage, transport, and setup
  • Safety yellow powder coat finish
  • Includes 304 stainless steel block anchor and zinc-plated Grade 5 hardware
  • Rated capacity of 450 lbs
  • Meets all applicable OSHA, ANSI, and CSA standards